Anti-IL17A Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 41809]

Supplier: R & D Systems
IC317N
102884-474EA 584.25 USD
102884-474
Anti-IL17A Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 41809]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 41809
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR