Anti-IL17A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 41809]

Supplier: R & D Systems
IC317A
102884-470EA 577.02 USD
102884-470
Anti-IL17A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 41809]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 41809
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR