Anti-IL17A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 41802]

Supplier: R & D Systems
IC3171G-100 IC3171G-025
102884-462EA 300.8 USD
102884-462 102884-464
Anti-IL17A Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 41802]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 41802
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR