Anti-IL17A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 41802]

Supplier: R & D Systems
IC3171A-025 IC3171A-100
102884-456EA 561.11 USD
102884-456 102884-454
Anti-IL17A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 41802]
Antibodies
Type: Primary
Antigen: IL17A
Clonality: Monoclonal
Clone: 41802
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR