Anti-CK8 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: LP3K]

Supplier: R & D Systems
IC3165G
102884-452EA 561.11 USD
102884-452
Anti-CK8 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: LP3K]
Antibodies
Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: LP3K
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR