Anti-IL32 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 373821]

Supplier: R & D Systems
IC30402A
102884-198EA 582.8 USD
102884-198
Anti-IL32 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 373821]
Antibodies
Type: Primary
Antigen: IL32
Clonality: Monoclonal
Clone: 373821
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR