Anti-ZBTB16 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6318100]

Supplier: R & D Systems
IC2944A
102884-190EA 594.37 USD
102884-190
Anti-ZBTB16 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 6318100]
Antibodies
Type: Primary
Antigen: ZBTB16
Clonality: Monoclonal
Clone: 6318100
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR