Anti-VEGFA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 23410]

Supplier: R & D Systems
IC2931G
102884-188EA 543.75 USD
102884-188
Anti-VEGFA Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 23410]
Antibodies
Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 23410
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR