Anti-STAT2 Rabbit Polyclonal Antibody (Fluorescein)

Supplier: R & D Systems
IC2890F
220018-372EA 420.83 USD
220018-372
Anti-STAT2 Rabbit Polyclonal Antibody (Fluorescein)
Antibodies
Type: Primary
Antigen: STAT2
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR