Anti-CCL5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 21445]

Supplier: R & D Systems
IC278A
220018-360EA 552.43 USD
220018-360
Anti-CCL5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 21445]
Antibodies
Type: Primary
Antigen: CCL5
Clonality: Monoclonal
Clone: 21445
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR