Anti-NES Mouse Monoclonal Antibody (PE) [clone: 307501]

Supplier: R & D Systems
IC2736P
220018-354EA 582.8 USD
220018-354
Anti-NES Mouse Monoclonal Antibody (PE) [clone: 307501]
Antibodies
Type: Primary
Antigen Symbol: NES
Antigen Name: nestin
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 307501
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR