Anti-CCL4 Mouse Monoclonal Antibody (Fluorescein) [clone: 24006]

Supplier: R & D Systems
IC271F
220018-352EA 420.83 USD
220018-352
Anti-CCL4 Mouse Monoclonal Antibody (Fluorescein) [clone: 24006]
Antibodies
Type: Primary
Antigen: CCL4
Clonality: Monoclonal
Clone: 24006
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR