Anti-CCL3 Mouse Monoclonal Antibody (Fluorescein) [clone: 93342]

Supplier: R & D Systems
IC2701F
220018-349EA 389.02 USD
220018-349
Anti-CCL3 Mouse Monoclonal Antibody (Fluorescein) [clone: 93342]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 93342
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR