Anti-CCL3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 93342]

Supplier: R & D Systems
IC2701A
220018-348EA 577.02 USD
220018-348
Anti-CCL3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 93342]
Antibodies
Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 93342
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR