Anti-GATA3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 291106]

Supplier: R & D Systems
IC26051P
102884-160EA 513.38 USD
102884-160
Anti-GATA3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 291106]
Antibodies
Type: Primary
Antigen Symbol: GATA3
Antigen Name: GATA binding protein 3
Antigen Synonyms: HDR, HDRS
Clonality: Monoclonal
Clone: 291106
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR