Anti-HBEGF Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 125923]

Supplier: R & D Systems
IC259G
102884-156EA 577.02 USD
102884-156
Anti-HBEGF Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 125923]
Antibodies
Type: Primary
Antigen Symbol: HBEGF
Antigen Name: heparin-binding EGF-like growth factor
Antigen Synonyms: DTR, DTS, DTSF, HEGFL
Clonality: Monoclonal
Clone: 125923
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR