Anti-LILRA3 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
IC2574G
102884-152EA 513.38 USD
102884-152
Anti-LILRA3 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: LILRA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR