Anti-IL27 Mouse Monoclonal Antibody (Fluorescein) [clone: 307426]

Supplier: R & D Systems
IC25261F
220018-341EA 420.83 USD
220018-341
Anti-IL27 Mouse Monoclonal Antibody (Fluorescein) [clone: 307426]
Antibodies
Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 307426
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR