Anti-BDNF Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 35909]

Supplier: R & D Systems
IC2481P
220018-337EA 513.38 USD
220018-337
Anti-BDNF Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 35909]
Antibodies
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone: 35909
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR