Anti-IL15 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 34559]

Supplier: R & D Systems
IC2471P
220018-335EA 582.8 USD
220018-335
Anti-IL15 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 34559]
Antibodies
Type: Primary
Antigen: IL15
Clonality: Monoclonal
Clone: 34559
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR