Anti-IL15 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 34559]

Supplier: R & D Systems
IC2471C
102884-148EA 594.37 USD
102884-148
Anti-IL15 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 34559]
Antibodies
Type: Primary
Antigen: IL15
Clonality: Monoclonal
Clone: 34559
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR