Anti-PAX3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 274212]

Supplier: R & D Systems
IC2457P
220018-333EA 513.38 USD
220018-333
Anti-PAX3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 274212]
Antibodies
Type: Primary
Antigen: PAX3
Clonality: Monoclonal
Clone: 274212
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR