Anti-PAX3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 274212]

Supplier: R & D Systems
IC2457A
220018-332EA 577.02 USD
220018-332
Anti-PAX3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 274212]
Antibodies
Type: Primary
Antigen: PAX3
Clonality: Monoclonal
Clone: 274212
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR