Anti-PDX1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 267712]

Supplier: R & D Systems
IC2419A
220018-330EA 577.02 USD
220018-330
Anti-PDX1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 267712]
Antibodies
Type: Primary
Antigen: PDX1
Clonality: Monoclonal
Clone: 267712
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR