Anti-IL22BP Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
IC2376A
220018-325EA 561.11 USD
220018-325
Anti-IL22BP Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: IL22BP
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR