Anti-OLIG1/2/3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 257224]

Supplier: R & D Systems
IC2230P
220018-317EA 543.75 USD
220018-317
Anti-OLIG1/2/3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 257224]
Antibodies
Type: Primary
Antigen: OLIG1/2/3
Clonality: Monoclonal
Clone: 257224
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR