Anti-IL12A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27537]

Supplier: R & D Systems
IC2191P
220018-316EA 582.8 USD
220018-316
Anti-IL12A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 27537]
Antibodies
Type: Primary
Antigen: IL12A
Clonality: Monoclonal
Clone: 27537
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR