Anti-STAT5A Mouse Monoclonal Antibody (Fluorescein) [clone: 251610]

Supplier: R & D Systems
IC21741F
220018-310EA 420.83 USD
220018-310
Anti-STAT5A Mouse Monoclonal Antibody (Fluorescein) [clone: 251610]
Antibodies
Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 251610
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR