Anti-STAT6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 253906]

Supplier: R & D Systems
IC2167A-100 IC2167A-025
102884-438EA 594.37 USD
102884-438 102884-436
Anti-STAT6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 253906]
Antibodies
Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: 253906
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR