Anti-CSF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26786]

Supplier: R & D Systems
IC2161P
220018-304EA 513.38 USD
220018-304
Anti-CSF1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 26786]
Antibodies
Type: Primary
Antigen: CSF1
Clonality: Monoclonal
Clone: 26786
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR