Anti-CSF3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5008]

Supplier: R & D Systems
IC2141P
220018-302EA 501.82 USD
220018-302
Anti-CSF3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5008]
Antibodies
Type: Primary
Antigen: CSF3
Clonality: Monoclonal
Clone: 5008
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR