Anti-IL13 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 32007]

Supplier: R & D Systems
IC2131P
220018-300EA 561.11 USD
220018-300
Anti-IL13 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 32007]
Antibodies
Type: Primary
Antigen: IL13
Clonality: Monoclonal
Clone: 32007
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR