Anti-TNF Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6402]

Supplier: R & D Systems
IC210P
220018-297EA 561.11 USD
220018-297
Anti-TNF Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6402]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 6402
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR