Anti-TNF Mouse Monoclonal Antibody (Fluorescein) [clone: 6402]

Supplier: R & D Systems
IC210F
220018-296EA 480.12 USD
220018-296
Anti-TNF Mouse Monoclonal Antibody (Fluorescein) [clone: 6402]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 6402
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR