Anti-VIM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 280618]

Supplier: R & D Systems
IC2105P
220018-295EA 561.11 USD
220018-295
Anti-VIM Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 280618]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280618
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR