Anti-VIM Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 280618]

Supplier: R & D Systems
IC2105G
102884-432EA 600.15 USD
102884-432
Anti-VIM Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 280618]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280618
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR