Anti-VIM Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 280618]

Supplier: R & D Systems
IC2105A
102884-430EA 633.42 USD
102884-430
Anti-VIM Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 280618]
Antibodies
Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: 280618
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR