Anti-IL9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 623153]

Supplier: R & D Systems
IC209A
102884-428EA 577.02 USD
102884-428
Anti-IL9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 623153]
Antibodies
Type: Primary
Antigen: IL9
Clonality: Monoclonal
Clone: 623153
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR