Anti-SMAD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 253343]

Supplier: R & D Systems
IC2097P
220018-294EA 513.38 USD
220018-294
Anti-SMAD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 253343]
Antibodies
Type: Primary
Antigen: SMAD4
Clonality: Monoclonal
Clone: 253343
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR