Anti-CXCL8 Mouse Monoclonal Antibody (Fluorescein) [clone: 6217]

Supplier: R & D Systems
IC208F
220018-292EA 404.92 USD
220018-292
Anti-CXCL8 Mouse Monoclonal Antibody (Fluorescein) [clone: 6217]
Antibodies
Type: Primary
Antigen: CXCL8
Clonality: Monoclonal
Clone: 6217
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR