Anti-T Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
IC2085G
102884-426EA 600.15 USD
102884-426
Anti-T Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen Symbol: T
Antigen Name: T brachyury transcription factor
Antigen Synonyms: SAVA, TFT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR