Anti-IL6 Mouse Monoclonal Antibody (Fluorescein) [clone: 1936]

Supplier: R & D Systems
IC206F
220018-289EA 438.18 USD
220018-289
Anti-IL6 Mouse Monoclonal Antibody (Fluorescein) [clone: 1936]
Antibodies
Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: 1936
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR