Anti-AKT (pan specific) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28146]

Supplier: R & D Systems
IC2055P
220018-285EA 561.11 USD
220018-285
Anti-AKT (pan specific) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 28146]
Antibodies
Type: Primary
Antigen: AKT (pan specific)
Clonality: Monoclonal
Clone: 28146
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR