Anti-AKT (pan specific) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 28146]

Supplier: R & D Systems
IC2055A
220018-283EA 600.15 USD
220018-283
Anti-AKT (pan specific) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 28146]
Antibodies
Type: Primary
Antigen: AKT (pan specific)
Clonality: Monoclonal
Clone: 28146
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR