Anti-IL4 Mouse Monoclonal Antibody (Fluorescein) [clone: 3007]

Supplier: R & D Systems
IC204F-100 IC204F-025
102884-416EA 513.38 USD
102884-416 102884-414
Anti-IL4 Mouse Monoclonal Antibody (Fluorescein) [clone: 3007]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 3007
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR