Anti-GATA2 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
IC2046P
220018-280EA 527.85 USD
220018-280
Anti-GATA2 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen Symbol: GATA2
Antigen Name: GATA binding protein 2
Antigen Synonyms: DCML, IMD21, MONOMAC, NFE1B
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR