Anti-CD68 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 298807]

Supplier: R & D Systems
IC20401P
220018-278EA 485.91 USD
220018-278
Anti-CD68 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 298807]
Antibodies
Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 298807
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR