Anti-IL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5334]

Supplier: R & D Systems
IC202P
220018-275EA 543.75 USD
220018-275
Anti-IL2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5334]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 5334
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR