Anti-SOX2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 245610]

Supplier: R & D Systems
IC2018G
102884-412EA 543.75 USD
102884-412
Anti-SOX2 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 245610]
Antibodies
Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: 245610
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR