Anti-SOX2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245610]

Supplier: R & D Systems
IC2018A
220018-267EA 594.37 USD
220018-267
Anti-SOX2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 245610]
Antibodies
Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: 245610
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR